Learn More
BIOHIPPO Research Grade Anti-Human APP/Amyloid beta Biosimilar Antibody (Merck anti-Amyloid beta 19.3), 100 µg
SDP
Supplier:  BIOHIPPO BHA21413505
Encompass_Preferred
APP/Amyloid beta is a research-grade biosimilar antibody. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.
Catalog No. 50-008-60278