Learn More
BIOHIPPO Research Grade Anti-Human CD200R1/OX2R Biosimilar Antibody (I-4P), 100 µg
SDP
Supplier:  BIOHIPPO BHA21413794
Encompass_Preferred
CD200R1/OX2R is a research-grade biosimilar antibody. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.
Catalog No. 50-008-60427