Learn More
BIOHIPPO Research Grade Anti-Human CD340/ERBB2/HER2/NEU Biosimilar Antibody (ALT-P7), 100 µg
Supplier: BIOHIPPO BHA21414411
ALT-P7 is a research-grade biosimilar antibody targeting CD340/ERBB2/HER2/NEU. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.