Learn More
BIOHIPPO Research Grade Anti-Human CD54/ICAM1 Biosimilar Antibody (MSH-TP15e), 100 µg
SDP
Supplier:  BIOHIPPO BHA21414347
Encompass_Preferred
MSH-TP15e is a research-grade biosimilar antibody targeting CD54/ICAM1. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.
Catalog No. 50-008-59361