Learn More
BIOHIPPO Research Grade Anti-Human FGF21 Biosimilar Antibody (CVX-343), 100 µg
SDP
Supplier:  BIOHIPPO BHA21414635
Encompass_Preferred
CVX-343 is a research-grade biosimilar antibody targeting FGF21. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.
Catalog No. 50-008-58723