Learn More
Cell Sciences Inc Elranatamab Research Grade BiTE Antibody, Bispecific T-Cell Engager, High Purity >99%, 1 mg
Supplier:  Cell Sciences Inc DHF92406B
Encompass
Elranatamab is a research-grade bispecific monoclonal antibody that targets BCMA and CD3, enabling T-cell-mediated destruction of myeloma cells. It offers >99% purity, making it ideal for high-quality research in oncology and immunology. This liquid formulation is stable, requires storage at -20°C for 12 months, and ships with dry ice, ensuring integrity for sensitive experiments.
Catalog No. NC2735810