Learn More
BIOHIPPO Research Grade Zovostotug Biosimilar Antibody, 100 µg
SDP
Supplier:  BIOHIPPO BHA21413838
Encompass_Preferred
Zovostotug is a research-grade biosimilar antibody. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.
Catalog No. 50-008-59282