Learn More
Selleck Chemical LLC Reserpine 50mg 50-55-5
SDP
Supplier:  Selleck Chemical LLC S160150MG
Encompass_Preferred
Reserpine is an inhibitor of multidrug efflux pumps, used as an antipsychotic and antihypertensive drug. Reserpine is an inhibitor of the vesicular monoamine transporter 2 (VMAT2). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8110