Learn More
Medchemexpress LLC Reserpine-d9 | 84759-11-5 | 617.73 | 100 UG
SDP
Supplier:  Medchemexpress LLC HYN0480S100UG
Encompass_Preferred
Reserpine-d9 is the deuterium-labeled form of Reserpine, an inhibitor of the vesicular monoamine transporter 2 (VMAT2). Deuterium-labeled compounds are valuable as tracers and internal standards for quantitative analysis in various analytical methods. The incorporation of stable heavy isotopes like deuterium into drug molecules can influence their pharmacokinetic and metabolic profiles, making them useful in drug development processes.
  • Can be used as a tracer.
  • Can serve as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes like deuterium are incorporated into drug molecules for quantitation during drug development.
  • Deuteration has the potential to impact pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-90128