Learn More
Sigma Aldrich Fine Chemicals Biosciences Reserpine, 50-55-5, MFCD00005091, 1 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 835801G
Encompass
Empirical Formula (Hill Notation): C33H40N2O9, Molecular Weight: 608.68, crystallized, ≥99.0% (HPLC), mp: ~265 °C, Synonym: (3β, 16β, 17α, 18β, 20α)-11,17-Dimethoxy-18-[(3,4,5-trimethoxybenzoyl)oxy]yohimban-16-carboxylic acid methyl ester.
Catalog No. NC0798223