Learn More
VEOLIA WTS RESERVOIR
Supplier:  VEOLIA WTS CAPF 9015002
Encompass
Low capacity oxidizer reagent cartridge for Sievers 900 and 800 Series. Replace after 3 months of use.
Catalog No. NC9851163