Learn More
Wilmad Labglass RESERVOIR CHROM 500ML 35/20
Supplier:  Wilmad Labglass LG4525102
Encompass_Preferred
Chromatography Reservoir, Spherical Joint. Capacity: 500mL, Joint Size: 35/20. For containment of mobile phase when larger volumes are required to elute from LG-4530 and LG-4535 chromatography columns
Catalog No. 50-804-180