Learn More
Medchemexpress LLC Resiquimod | 144875-48-9 | MFCD00937759 | >98.0% | 314.39 g/mol | C17H22N4O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY137405MG
Encompass_Preferred
Resiquimod is a Toll-like receptor 7 and 8 (TLR7/TLR8) agonist used in research to stimulate innate immune responses. It promotes production of cytokines such as TNF-α, IL-6, and IFN-α and is commonly used in studies of immunomodulation, vaccine adjuvants, and infection models.
  • Activates TLR7 and TLR8 to induce innate immune signaling.
  • Induces cytokines such as TNF-α, IL-6, and IFN-α.
  • Suitable for in vitro and in vivo immunology research.
  • Available in small research quantities for analytical and experimental use.
  • Stable as a powder when stored at recommended temperatures.
Catalog No. 50-002-21799