Learn More
Medchemexpress LLC Resiquimod-d5 | 2252319-44-9 | 99.8% | 319.41 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13740S10MM/1ML
Encompass_Preferred
Resiquimod-d5 is a deuterium-labeled variant of Resiquimod, which functions as a Toll-like receptor 7 and 8 (TLR7/TLR8) agonist. This compound is known to induce the upregulation of various cytokines, including TNF-α, IL-6, and IFN-α, making it valuable for immunological research.
  • Can be utilized as a tracer in various experimental setups.
  • Suitable for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS due to its stable isotope labeling.
  • Deuteration can influence the pharmacokinetic and metabolic profiles of drugs, offering insights into drug development.
  • Induces the upregulation of key cytokines like TNF-α, IL-6, and IFN-α through TLR7/TLR8 agonism.
Catalog No. 50-003-49759