Learn More
Medchemexpress LLC Resiquimod (R848; S28463) | 144875-48-9 | 314.38 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13740G50MG
Encompass_Preferred
Resiquimod (R848) (GMP) is a version of Resiquimod manufactured following Good Manufacturing Practice (GMP) guidelines, making it suitable as an auxiliary reagent for cell therapy manufacturing. This compound functions as an agonist for Toll-like receptor 7 and 8 (TLR7/TLR8), demonstrating various cellular effects.
  • Can induce human mMDSC to mature into inflammatory macrophages.
  • Can produce IL-6 and IL-12 in human mMDSC.
  • Can transdifferentiate memory B cells into IgG producing plasma cells.
  • Can induce the differentiation of MDSCs into mature myeloid cells.
Catalog No. 50-003-49758