Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer General Resistance DA-52-3X Decade Box0.01%Acc Resistance 5 Decades

Supplier:  Cole-Parmer 2001889

Encompass_Preferred

RDS SERIES

  • Accuracies 0.1% or 0.025%
  • 4, 5, 6 or 7 decades
  • "Top of the line" GR performance
  • Rotary switches, gold-plated contacts, low contact R
  • Minimum Ohms = 0
  • OEM models available on special order
DA SERIES
  • Accuracies .01% or 0.1%
  • 3, 4,5,6 or 7 decades
  • Less than one-half the cost of RDS units
  • Easy to use, economical thumbwheel switches
  • Two mechanical configurations
    Note: GR Decade Boxes as RTD Simulators When the ultimate in accuracy is required, GR's RTD-100X or RTD-100RTD should be used Thes

  • Catalog No. 50-632-282


    May include imposed supplier surcharges.
    Only null left
    Add to Cart

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.