Learn More
GA International XyliTUFF™ – Extended Exposure Xylene & Chemical-Resistant Labels
Supplier:  GA International FTT233C13WH
Encompass_Preferred
These labels are specifically designed for use in histology and cytology labs, ideally suited for long-term identification of microscope slides, slide boxes, and other containers. Resistant to extended chemical exposure, the labels withstand common organic solvents, industrial thinners, flux, oils, grease, degreasers, gasoline, as well as other chemicals and solvents. Intended for use on flat surfaces, these solvent-resistant labels along with their printout can withstand immersion in xylene for up to 15 hours (on glass surfaces). Compatible with all major commercially available thermal-transfer barcode printers, these chemical resistant histology and industry-grade labels can be printed with alphanumeric text, 1D and 2D barcodes and serialized numbers.
Catalog No. NC1643976