Learn More
AdipoGen Resistomycin
SDP
Supplier:  AdipoGen BVT0036M001
Encompass_Preferred
Chemical. CAS 20004-62-0. Formula C22H16O6. MW 376.4. Isolated from Streptomyces sp. Antibacterial Gram-positive and mycobacteria. RNA polymerase inhibitor. Apoptosis inducer.
Catalog No. 50-256-2131