Learn More
Medchemexpress LLC Resolvin D1-d5 | 1881277-32-2 | 99.2% | 381.52 | C22H27D5O5 | 10UG
SDP
Supplier:  Medchemexpress LLC HY125527S10UG
Encompass_Preferred
Resolvin D1-d5 is a deuterium-labeled internal standard of Resolvin D1 supplied for analytical and mass-spectrometry workflows. It is intended for research use and is used as an internal standard for accurate quantification of RvD1 in LC/GC-MS procedures. The product is provided in small vials (including a 10 μg ethanol solution) and has high reported purity.
Catalog No. 50-002-35098