Learn More
Cayman Chemical ResolvIn D1 methyl ester 10UG
SDP
Supplier:  Cayman Chemical 900181510 UG
Encompass_Preferred
A methyl ester version of the free acid that may act as a lipophilic prodrug form that will alter its distribution and pharmacokinetic properties
Catalog No. 50-289-3726