Learn More
Medchemexpress LLC Resolvin D2-d5 | 1881277-33-3 | 99.0% | 10 UG
SDP
Supplier:  Medchemexpress LLC HY121636S10UG
Encompass_Preferred
Resolvin D2-d5 is a deuterium-labeled internal standard of Resolvin D2 used as a tracer and quantitative internal standard for NMR, GC-MS, and LC-MS analyses. Supplied as a 10 μg solution in ethanol with high isotopic and chemical purity, it is intended for analytical quantification and tracer studies.
  • Deuterium-labeled tracer for accurate mass spectrometric quantification.
  • Supplied as a ready-to-use solution to simplify sample preparation.
  • High chemical and isotopic purity for reliable analytical results.
  • Suitable for NMR, GC-MS, and LC-MS as an internal standard.
  • Stable when stored at -20°C for up to two years.
Catalog No. 50-002-30438