Learn More
Cayman Chemical ResolvIn D2 methyl ester 100UG
SDP
Supplier:  Cayman Chemical 9001497100 UG
Encompass_Preferred
A methyl ester version of RvD2 which may act as a lipophilic prodrug form that will alter its distribution and pharmacokinetic properties
Catalog No. 50-289-6045