Learn More
Cayman Chemical ResolvIn D3 25ug
SDP
Supplier:  Cayman Chemical 1383425 UG
Encompass_Preferred
A DHA-derived product first identified in mouse inflammatory exudates; reduces neutrophil infiltration in vivo in both mouse peritonitis and dermal inflammation; enhances macrophage phagocytosis and efferocytosis
Catalog No. 50-295-5232