Learn More
Cayman Chemical ResolvIn D3 methyl ester 50UG
SDP
Supplier:  Cayman Chemical 900268650 UG
Encompass_Preferred
A methyl ester form of resolvin D3; may act as a lipophilic prodrug form that will alter its distribution and pharmacokinetic properties
Catalog No. 50-289-2925