Learn More
Cayman Chemical ResorcInolnaphthaleIn 5mg
SDP
Supplier:  Cayman Chemical 130825 MG
Encompass_Preferred
Increases the activity of angiotensin-converting enzyme 2 (ACE2) in vitro (EC50 = 19.5 μM)
Catalog No. 50-295-5932