Learn More
Medchemexpress LLC Resorufin | 635-78-9 | MFCD00128991 | 98.9% | 213.19 g/mol | C12H7NO3 | 50MG
SDP
Supplier:  Medchemexpress LLC HY12353350MG
Encompass_Preferred
Resorufin is a highly fluorescent red-pink dye and probe used in biochemical and cell-based assays to detect reactive oxygen and nitrogen species and to report enzymatic activity. It is water-soluble, exhibits red-shifted absorption and emission, and commonly serves as the fluorescent product in assay chemistries.
Catalog No. 50-002-38476