Learn More
Medchemexpress LLC Resorufin acetate | 1152-14-3 | MFCD00037659 | ≥98.0% | 255.22 g/mol | C14H9NO4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11826210MG
Encompass_Preferred
Resorufin acetate is a fluorogenic probe (CAS 1152-14-3) used as a substrate in enzyme and cell-based assays. Esterase-mediated hydrolysis produces resorufin, a red fluorescent product, enabling sensitive fluorometric detection. Supplied as a solid in small milligram quantities for research use.
  • Fluorogenic substrate for esterase and aldehyde dehydrogenase (ALDH1A1) assays.
  • Produces a red fluorescent signal (resorufin) upon hydrolysis for quantitative detection.
  • High purity (≥98%) suitable for sensitive assays.
  • Soluble in DMSO and compatible with common assay buffers.
  • Stable when stored at -20°C to preserve activity.
Catalog No. 50-002-26945