Learn More
AdipoGen Resorufin ethyl ether
SDP
Supplier:  AdipoGen CDXR0013M100
Encompass_Preferred
Chemical. CAS 5725-91-7. Formula C14H11NO3. MW 241.24. Synthetic. Dealkylase substrate for the microfluorimetric analysis of microsomal cytochrome P-450.
Catalog No. 50-256-0146