Learn More
AdipoGen Resorufin methyl ether
SDP
Supplier:  AdipoGen CDXR0012M001
Encompass_Preferred
Chemical. CAS 5725-89-3. Formula C13H9NO3. MW 227.22. Synthetic. Substrate for Cytochrome P-450. Used for various investigations including the differentiatiation of cytochrome P-450 isozymes.
Catalog No. 50-256-0151