Learn More
Medchemexpress LLC Resorufin methyl ether | 5725-89-3 | 99.8% | 5 MG
Supplier: Medchemexpress LLC HYD01445MG
Resorufin methyl ether (Methoxyresorufin) is a cytochrome P450 fluorometric substrate. It is a relatively specific substrate for CYP1A2 activity in rodents.
- Cytochrome P450 fluorometric substrate
- Relatively specific substrate for CYP1A2 activity in rodents