Learn More
Medchemexpress LLC Resorufin sodium salt | 34994-50-8 | MFCD00005037 | 99.6% | 235.17 g/mol | C12H6NNaO3 | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY123533A10MM 1ML
Encompass_Preferred
Resorufin sodium salt is a water-soluble fluorescent dye and redox indicator used in fluorescence-based assays, reactive oxygen/nitrogen species detection, and as a calibration standard for resorufin-derived substrates. It is available as a solid and as pre-prepared 10 mM solutions for convenient use in biochemical workflows.
  • Highly fluorescent red-emitting dye suitable for fluorescence assays.
  • Water-soluble sodium salt; available as solid and 10 mM solutions.
  • Useful as a redox indicator and probe for ROS/RNS detection.
  • Suitable as a calibration standard for resorufin-derived enzyme substrates.
  • High purity (99.6%) with molecular weight 235.17 g/mol.
Catalog No. 50-002-38586