Learn More
Sigma Aldrich Fine Chemicals Biosciences RESOURCETM PHE Cytiva 17 1
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17118601
Encompass
RESOURCE(TM) PHE are prepacked SOURCE(TM) 15 PHE (phenyl) columns for purification of proteins and peptides with hydrophobic interaction chromatography (HIC). For fast high-reSOLUTIONon purifications RESOURCE(TM) columns are excellent choice for polishing and removal of trace impurities.
Catalog No. 50-290-8036