Learn More
MSC North Facepiece Respirator Cleaning Wipes, 5 x 7 in, 100/Pack
Supplier:  MSC 82727280
Encompass
These facepiece respirator cleaning wipes are designed for effective cleaning and refreshing of respirator equipment. Formulated with Propan-2-ol and water, they are suitable for maintaining hygiene and performance of respirator facepieces.
  • Individually wrapped for convenience and to prevent contamination
  • Made from cloth material for gentle yet effective cleaning
  • White color
  • Compatible with both full mask and half mask respirators
  • Contains 70% Propan-2-ol and 30% water
Catalog No. 50-237-0295