Learn More
Essendant RESPIRATOR42 CFR 84 MED
Supplier:  Essendant MMM6291
Encompass_Preferred
Here s the solution for workers who frequently need a respirator for job after job It s reusablejust replace cartridges and filters when used to capacity Comfortable soft and lightweight with easy-to-adjust headstraps Suitable for use against a variety of gases vapors and particulate hazards Filters and cartridges sold separately Respirator Type- Half-Face Mask Rating- P100 Material(s)- Thermoplastic Elastomer (TPE) Valved- Yes
Catalog No. 50-000-35811