Learn More
Essendant RESPIRATOR7700HALFLG
Supplier:  Essendant NSP770030L
Encompass_Preferred
Comfortable half masks are suitable for multiple work environments and can be adapted to PAPR or Supplied Air systems Because of low dead air space within the mask less exhaled air is recycled Strategic placement of the exhalation valve means that more breath leaves the mask swiftly creating a cooler environment for the wearer 7700 series half masks are 100pct silicone making them an ideal solution for workers with latex allergies Dual-elastic headstraps are carefully engineered bottom straps have more elasticity than top straps to create a secure non-slip fit Wide contoured sealing area provides an improved fit for a variety of facial types Respirator Type- Half-Face Mask Material(s)- Silicone Valved- Yes Size- Large
Catalog No. 50-000-31543