Learn More
Essendant RESPIRATORN95 VFLEXWH
Supplier:  Essendant MMM9105
Encompass_Preferred
Economical disposable N95 particulate respirator is designed to help provide quality reliable worker respiratory protection for certain non-oil based particles It has a unique folded design with 3M proprietary filter media and V-shaped pleats that expand to help make breathing easier In addition the V-shaped pleats flex with mouth movement for easier talking The respirator features uniquely designed tabs for positioning the respirator on the face and embossed front panel to help the respirator retain its shape Flat-fold design allows for convenient storage prior to use Adjustable noseclip helps provide a custom fit and secure seal Uniquely designed tabs for respirator positioning on the face Spacious inside with comfortable inner layer Compatible with a variety of eyewear and hearing protection Respirator Type- Half-Face Mask Rating- Unvalved N95 Airborne Contaminant Resistance- Non-Oil Based Particulates Material(s)- Advanced Electrostatic Media
Catalog No. 50-000-31179