Learn More
Essendant RESPIRATORP95 MAINT.FREE
Supplier:  Essendant MMM8577
Encompass_Preferred
Respirator provides P95 level protection against oil and non-oil based aerosols Added carbon-layering to the filter helps minimize organic vapor bleed-through Lightweight design and adjustable M-noseclip enhance comfort levels particularly over extended use periods Ideal for hot and humid work environments Nuisance Level Organic Vapor Relief Respirator Type- Half-Face Mask Rating- P95 Valved- Yes Size- One Size Fits All
Catalog No. 50-000-35939