Learn More
Essendant RESPIRATORPNT SPRY and PST
Supplier:  Essendant MMM53P71
Encompass_Preferred
The 3M 5000 Series Half Facepiece Disposable Respirator Assembly with Organic Vapor and P95 Particulate Filters Respiratory Protection (51P71 52P71 53P71) helps provide protection against certain airborne contaminants This respirator is NIOSH approved for respiratory protection against certain organic vapors and particulates up to ten times the permissible exposure limit (PEL) Half Facepiece respirator may be used in a variety of workplace applications including spray painting pesticides agriculture construction and transportation This disposable respirator assembly offers greater convenience by reducing the need for long-term maintenance and is an ideal choice for intermittent use NIOSH color code of Black Respirator Type- Half-Face Mask Rating- P95 Airborne Contaminant Resistance- Organic Vapors Material(s)- Thermoplastic Elastomer (TPE)
Catalog No. 50-000-32853