Learn More
Essendant RESPIRATORR95 W/LVL OV
Supplier:  Essendant MMM8247
Encompass_Preferred
Chemical absorbent removes odors Lightweight construction and contoured fit enhance comfort and wear-time Easy to breathe through even without an exhalation valve Provides relief from latex paint odor unpleasant smog levels and non-hazardous household odors Do not use for asbestos gasses vapors above the government permissible exposure limits or sandblasting Respirator Type- Half-Face Mask Rating- N95 Size- One Size Fits All Quantity- 20/Box
Catalog No. 50-000-31130