Learn More
Restek Corporation Restek Split Liners for PerkinElmer: Baffle Splitter
SDP
Supplier:  Restek Corporation 20737
Encompass_Preferred
Sleeve, Baffle Splitter; 3.5mm I.D.; For Perkinelmer GCs; Length: 100mm; 5.0mm O.D.; Pack of 5
Catalog No. 06-717-879