Learn More
Restek Corporation Restek Splitless Liners for Varian 1075/1077 GCs
SDP
Supplier:  Restek Corporation 20897
Encompass_Preferred
Sleeve, Cyclo Double Gooseneck; 4mm I.D.; For Varian 1075/1077 GCs; Length: 74mm; 6.3mm O.D.
Catalog No. 06-711-187