Learn More
Bioworld Resurrection Media Concentrate (100X), 50 mL
Supplier:  Bioworld 306200711
Encompass_Preferred
Resurrection Media ConcentrateEnsures the survival of bacterial strains and clones during prolonged storage at 4 °C to -80 °C100X concentration
Catalog No. 50-254-363