Learn More
Bioworld Resurrection Media Pack, 100 L
Supplier:  Bioworld 306200722
Encompass_Preferred
Resurrection Media PackEnsures the survival of bacterial strains and clones during prolonged storage at 4 °C to -80 °C Add 1 pack/L to any culture mediaSold 10 pack for 10x1L media.
Catalog No. 50-254-366