Learn More
Sino Biological Recombinant Human RET Protein (V738A, GST Tag), Active
Supplier:  Sino Biological R0212RG500
Encompass_Preferred
Recombinant human RET (V738A) (658-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3406