Learn More
Cayman Chemical RetapamulIn 250mg
SDP
Supplier:  Cayman Chemical 21528250 MG
Encompass_Preferred
A pleuromutilin antibiotic that inhibits bacterial protein synthesis by binding to bacterial ribosomes in the peptidyl transferase component of the 50S subunit and inhibiting peptide bond formation
Catalog No. 50-298-6500