Learn More
Medchemexpress LLC Retaspimycin hydrochloride | 857402-63-2 | 96.2% | 624.17 g/mol | C31H46ClN3O8 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1021010MG
Encompass_Preferred
Retaspimycin hydrochloride is a research-grade small-molecule inhibitor of heat shock protein 90 (Hsp90) and Grp94, used in biochemical and cellular studies to probe Hsp90-dependent pathways. It is supplied as the hydrochloride salt with a molecular weight of 624.17 g/mol and reported purity of 96.16%.
  • Potent Hsp90 inhibitor with reported EC50 ≈119 nM for Hsp90 and Grp94.
  • High purity suitable for in vitro biochemical and cellular assays.
  • Available in multiple solid and solution pack sizes for flexible experimental use.
  • Provided with datasheet, certificate of analysis, and safety data sheet for quality verification.
  • Hydrochloride salt form improves handling and solubility in polar solvents.
Catalog No. 50-002-47116