Learn More
Medchemexpress LLC Retaspimycin hydrochloride | 857402-63-2 | 96.2% | 624.17 g/mol | C31H46ClN3O8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY102105MG
Encompass_Preferred
Retaspimycin hydrochloride is the hydrochloride salt of a small-molecule inhibitor of heat-shock protein 90 (Hsp90) and the endoplasmic reticulum paralog Grp94. It is supplied for laboratory research use and has reported EC50 values of 119 nM for both Hsp90 and Grp94.
  • Potent inhibitor of Hsp90 and Grp94 (EC50 119 nM).
  • Hydrochloride salt form for improved handling and stability.
  • Molecular formula C31H46ClN3O8 and molecular weight 624.17 g/mol.
  • Supplied in small-mass quantities suitable for research (e.g., 5 mg).
  • Typical purity reported by supplier: 96.16% (LC/HPLC documentation available).
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-46940