Learn More
Medchemexpress LLC Retaspimycin hydrochloride | 857402-63-2 | 96.2% | 624.17 g/mol | C31H46ClN3O8 | 50 MG

Supplier: Medchemexpress LLC HY1021050MG
Retaspimycin hydrochloride is the hydrochloride salt of retaspimycin (IPI-504), a semi-synthetic geldanamycin analogue that potently inhibits heat shock protein 90 (Hsp90). It is supplied as a solid research reagent for in vitro and preclinical studies; supplier-reported purity is ~96.16% and molecular weight is 624.17 g/mol.
- Potent Hsp90 inhibitor, EC50 ~119 nM for Hsp90 and Grp94.
- Research-grade solid suitable for biochemical and cell-based assays.
- Supplier-reported purity 96.16% (typical lot specification).
- Molecular weight 624.17 g/mol.
- Recommended storage sealed, away from moisture at -20°C; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
- Available in small solid package sizes for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.