Learn More
Selleck Chemical LLC Retatrutide P1230-25MG
SDP
Supplier:  Selleck Chemical LLC P123025MG
Encompass_Preferred
Retatrutide (LY3437943) acts as a triple agonist peptide of the glucagon receptor (GCGR) glucagon-like peptide-1 receptor (GLP-1R) and glucosedependent insulinotropic polypeptide receptor (GIPR) Retatrutide binds to human GCGR GLP-1R and GIPR with EC50 values of 5 79 0 775 nM and 0 0643 respectively Retatrutide has the potential for use in research on obesity
Catalog No. 50-000-43643