Learn More
Electron Microscopy Sciences Reticle, 10 mm Scale With 100 Divisions
SDP
Supplier:  Electron Microscopy Sciences A910100
Encompass_Preferred
Reticles for use with our compound and inverted light microscopes. Reticle with 10 mm scale with 100 divisions for use with compound microscopes.
Catalog No. 50-342-66